The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029582.2 | NP_083858.1 | 10 - 11 | 2164 | 77 |
NM_134105.2 | NP_598866.1 | 3 - 4 | 407 | 77 | |
GenBank mRNA | AK139620.1 | - | 10 - 11 | 2247 | 77 |
AK155844.1 | - | 10 - 11 | 2218 | 77 | |
AK156154.1 | - | 10 - 11 | 2205 | 77 | |
AK157288.1 | - | 10 - 11 | 2138 | 77 | |
BC019564.1 | - | 3 - 4 | 432 | 77 | |
BC020363.1 | - | 9 - 10 | 1611 | 77 |