Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4f15
Entrez Gene ID
106648
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 15
Gene Aliases
AI787289, Cyp4f-15
Chromosome Location
Chr.17: 32685601 - 32703352 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134127.1 NP_598888.1 12 - 13 1574 79
GenBank mRNA AF233645.1 - 12 - 13 1574 79
BC021377.1 - 12 - 13 1544 79

View Full Product Details