Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Macrod1
Entrez Gene ID
107227
Gene Name
MACRO domain containing 1
Gene Aliases
AI604841, AW743046, D930010J01Rik, Lrp16
Chromosome Location
Chr.19: 7056715 - 7198062 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134147.4 NP_598908.2 8 - 9 1008 71
GenBank mRNA AI604841.1 - 3 - 4 71
AK052998.1 - 8 - 9 967 71
BC008653.1 - 8 - 9 733 71

View Full Product Details