The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134150.2 | NP_598911.1 | 1 - 2 | 84 | 93 |
GenBank mRNA | AK027996.1 | - | 1 - 2 | 71 | 93 |
AK087195.1 | - | 1 - 2 | 71 | 93 | |
AK140070.1 | - | 1 - 2 | 71 | 93 | |
AK145273.1 | - | 1 - 2 | 74 | 93 | |
AK145970.1 | - | 1 - 2 | 76 | 93 | |
BC022575.1 | - | 1 - 2 | 84 | 93 | |
BC054410.1 | - | 1 - 2 | 52 | 93 |