The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134248.2 | NP_599009.2 | 3 - 4 | 552 | 80 |
GenBank mRNA | AF399829.1 | - | 3 - 4 | 543 | 80 |