The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145823.2 | NP_665822.1 | 2 - 3 | 980 | 65 |
GenBank mRNA | AB077281.1 | - | 2 - 3 | 961 | 65 |
AB077282.1 | - | 2 - 3 | 258 | 65 | |
AK003844.1 | - | 1 - 2 | 228 | 65 | |
AK021206.1 | - | 2 - 3 | 193 | 65 | |
AK153625.1 | - | 2 - 3 | 981 | 65 | |
AK171340.1 | - | 2 - 3 | 1007 | 65 | |
AK172137.1 | - | 2 - 3 | 981 | 65 | |
BC082333.1 | - | 2 - 3 | 812 | 65 | |
BC108351.1 | - | 2 - 3 | 840 | 65 |