Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pitpnc1
Entrez Gene ID
71795
Gene Name
phosphatidylinositol transfer protein, cytoplasmic 1
Gene Aliases
1110020B03Rik, 5830436L09Rik, AI662802, AI851387, C330017I21Rik, Dnr411, RDGBB, RDGBB1, rdgB-beta
Chromosome Location
Chr.11: 107207892 - 107470892 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145823.2 NP_665822.1 9 - 10 1578 80
GenBank mRNA AB077281.1 - 9 - 10 1559 80
AK171918.1 - 9 - 10 1575 80
BC082333.1 - 9 - 10 1410 80
BC108351.1 - 9 - 10 1438 80

View Full Product Details