Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Las1l
Entrez Gene ID
76130
Gene Name
LAS1-like (S. cerevisiae)
Gene Aliases
1810030A06Rik, 5830482G23Rik
Chromosome Location
Chr.X: 95920045 - 95956974 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152822.3 NP_690035.2 4 - 5 597 69
GenBank mRNA AK018059.1 - 4 - 5 527 69
AK163919.1 - 3 - 4 2424 69
BC004735.1 - 3 - 4 189 69
BC032218.1 - 4 - 5 520 69
BC141155.1 - 4 - 5 534 69
BQ573371.1 - 3 - 4 177 69

View Full Product Details