The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009660.3 | NP_033790.3 | 1 - 2 | 162 | 73 |
GenBank mRNA | AK142371.1 | - | 1 - 2 | 163 | 73 |
BC056625.1 | - | 1 - 2 | 149 | 73 | |
BC081546.1 | - | 1 - 2 | 148 | 73 | |
CT010263.1 | - | 1 - 2 | 136 | 73 | |
U04331.1 | - | 1 - 2 | 136 | 73 |