Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Add3
Entrez Gene ID
27360
Gene Name
adducin 3 (gamma)
Gene Aliases
AI463285, R75380
Chromosome Location
Chr.19: 53140443 - 53247326 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164099.2 NP_001157571.1 3 - 4 622 73
NM_001164100.2 NP_001157572.1 3 - 4 684 73
NM_001164101.2 NP_001157573.1 3 - 4 622 73
NM_001277100.1 NP_001264029.1 3 - 4 684 73
NM_013758.4 NP_038786.2 4 - 5 434 73
GenBank mRNA AF100424.1 - 2 - 3 353 73
AF100425.1 - 2 - 3 353 73
AK019491.1 - 3 - 4 563 73
AK028481.1 - 3 - 4 527 73
AK035151.1 - 3 - 4 451 73
AK083939.1 - 3 - 4 525 73
AK144174.1 - 3 - 4 628 73
BC004032.1 - 3 - 4 401 73
BC029196.1 - 3 - 4 487 73
BC037116.1 - 4 - 5 445 73
BC060097.1 - 3 - 4 466 73

View Full Product Details