The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290807.1 | NP_001277736.1 | 5 - 6 | 473 | 84 |
NM_001290808.1 | NP_001277737.1 | 4 - 5 | 503 | 84 | |
NM_025475.3 | NP_079751.1 | 5 - 6 | 573 | 84 | |
GenBank mRNA | AK007106.1 | - | 5 - 6 | 475 | 84 |
AK010445.1 | - | 5 - 6 | 520 | 84 | |
AK012086.1 | - | 5 - 6 | 572 | 84 | |
AK012779.1 | - | 4 - 5 | 503 | 84 | |
AK012960.1 | - | 5 - 6 | 520 | 84 | |
AK132300.1 | - | 5 - 6 | 529 | 84 | |
BC086677.1 | - | 5 - 6 | 511 | 84 |