The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025731.2 | NP_080007.1 | 4 - 5 | 579 | 93 |
GenBank mRNA | AB261168.1 | - | 4 - 5 | 537 | 93 |
AB447517.1 | - | 4 - 5 | 537 | 93 | |
AK006527.1 | - | 4 - 5 | 544 | 93 | |
AK006790.1 | - | 4 - 5 | 544 | 93 | |
AK014971.1 | - | 4 - 5 | 580 | 93 | |
BC061008.1 | - | 4 - 5 | 559 | 93 |