Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc77
Entrez Gene ID
67200
Gene Name
coiled-coil domain containing 77
Gene Aliases
2400002C23Rik, 2700091N06Rik, AV168274
Chromosome Location
Chr.6: 120324322 - 120364369 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026028.5 NP_080304.1 5 - 6 675 78
GenBank mRNA AK010251.1 - 5 - 6 528 78
AK012598.1 - 5 - 6 567 78

View Full Product Details