The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027213.2 | NP_081489.2 | 3 - 4 | 270 | 108 |
GenBank mRNA | AK005232.1 | - | 3 - 4 | 315 | 108 |
AK089101.1 | - | 3 - 4 | 319 | 108 | |
AK171784.1 | - | 3 - 4 | 286 | 108 | |
BC013096.1 | - | 3 - 4 | 270 | 108 |