The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027332.3 | NP_081608.1 | 1 - 2 | 258 | 94 |
NM_027371.3 | NP_081647.2 | 1 - 2 | 258 | 94 | |
GenBank mRNA | AK008988.1 | - | 1 - 2 | 247 | 94 |
AK010079.1 | - | 1 - 2 | 255 | 94 | |
AK162073.1 | - | 1 - 2 | 261 | 94 | |
AK171035.1 | - | 1 - 2 | 239 | 94 | |
AK171079.1 | - | 1 - 2 | 247 | 94 | |
BC056165.1 | - | 1 - 2 | 209 | 94 | |
BC089369.1 | - | 1 - 2 | 243 | 94 |