The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027439.4 | NP_081715.1 | 3 - 4 | 445 | 64 |
GenBank mRNA | AB192471.2 | - | 3 - 4 | 321 | 64 |
AK017482.1 | - | 3 - 4 | 377 | 64 | |
AK029405.1 | - | 3 - 4 | 348 | 64 | |
AK076495.1 | - | 3 - 4 | 390 | 64 | |
AK151735.1 | - | 3 - 4 | 376 | 64 | |
AK168139.1 | - | 3 - 4 | 402 | 64 | |
BC014706.1 | - | 3 - 4 | 316 | 64 |