Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spata16
Entrez Gene ID
70862
Gene Name
spermatogenesis associated 16
Gene Aliases
4921511F01Rik, 4930503K02Rik, Nyd-sp12
Chromosome Location
Chr.3: 26637631 - 26983212 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027583.3 NP_081859.1 6 - 7 1210 71
NM_029150.4 NP_083426.1 6 - 7 1210 71
GenBank mRNA AK014869.1 - 6 - 7 1174 71
AK076628.1 - 6 - 7 1211 71
BC100443.1 - 6 - 7 1191 71
BC132205.1 - 6 - 7 1134 71
BC132539.1 - 6 - 7 1134 71

View Full Product Details