The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136237.1 | NP_001129709.1 | 6 - 7 | 1015 | 88 |
NM_028051.3 | NP_082327.2 | 5 - 6 | 1101 | 88 | |
NM_028092.3 | NP_082368.1 | 5 - 6 | 977 | 88 | |
GenBank mRNA | AK007473.1 | - | 5 - 6 | 1102 | 88 |
AK008448.1 | - | 5 - 6 | 978 | 88 | |
BC028990.1 | - | 6 - 7 | 976 | 88 |