The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028121.2 | NP_082397.1 | 5 - 6 | 899 | 86 |
GenBank mRNA | AK011434.1 | - | 5 - 6 | 864 | 86 |
AK037017.1 | - | - | 1585 | 86 | |
AK149942.1 | - | 5 - 6 | 871 | 86 | |
AK162169.1 | - | 5 - 6 | 852 | 86 | |
AK167634.1 | - | 5 - 6 | 900 | 86 | |
AK169590.1 | - | 5 - 6 | 863 | 86 | |
AK214218.1 | - | 1 - 2 | 115 | 86 | |
BC021526.1 | - | 5 - 6 | 886 | 86 | |
BC094657.1 | - | 4 - 5 | 3579 | 86 |