Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gcc1
Entrez Gene ID
74375
Gene Name
golgi coiled coil 1
Gene Aliases
4932417P04Rik
Chromosome Location
Chr.6: 28416603 - 28421724 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK016531.1 - - 295 81

View Full Product Details