The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168318.1 | NP_001161790.1 | 1 - 2 | 342 | 84 |
NM_028903.2 | NP_083179.2 | 1 - 2 | 342 | 84 | |
GenBank mRNA | AK016912.1 | - | 1 - 2 | 309 | 84 |
AK033154.1 | - | 1 - 2 | 309 | 84 | |
AK133176.1 | - | 1 - 2 | 343 | 84 | |
BC016096.1 | - | 1 - 2 | 318 | 84 | |
BC023907.1 | - | 1 - 2 | 258 | 84 | |
BC032159.1 | - | 1 - 2 | 298 | 84 |