Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scara5
Entrez Gene ID
71145
Gene Name
scavenger receptor class A, member 5 (putative)
Gene Aliases
4932433F15Rik, 4933425F03Rik, AV278087, Tesr
Chromosome Location
Chr.14: 65666403 - 65764826 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168318.1 NP_001161790.1 5 - 6 1344 82
NM_028903.2 NP_083179.2 5 - 6 1344 82
GenBank mRNA AK016543.1 - 5 - 6 1379 82
AK016912.1 - 5 - 6 1311 82
AK033154.1 - 5 - 6 1311 82
AK133176.1 - 5 - 6 1345 82
BC016096.1 - 5 - 6 1320 82
BC023907.1 - 5 - 6 1260 82
BC026758.1 - 2 - 3 370 82
BC032159.1 - 5 - 6 1300 82
DQ122126.1 - 4 - 5 994 82

View Full Product Details