Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rprd2
- Entrez Gene ID
-
75137
- Gene Name
- regulation of nuclear pre-mRNA domain containing 2
- Gene Aliases
-
2810036A19Rik, 4930535B03Rik, 6720469I21Rik, AL022841, AL022940, AU021304, BB077382, mKIAA0460
- Chromosome Location
-
Chr.3: 95759873 - 95819723 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK015972.1 |
- |
3 - 4 |
549 |
99 |
AK083674.1 |
- |
3 - 4 |
461 |
99 |
AK087157.1 |
- |
3 - 4 |
464 |
99 |
View Full Product Details