Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Armcx1
Entrez Gene ID
78248
Gene Name
armadillo repeat containing, X-linked 1
Gene Aliases
3010033I09Rik, ALEX1
Chromosome Location
Chr.X: 134717912 - 134721917 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030066.3 NP_084342.1 4 - 5 366 130
GenBank mRNA AK019405.1 - 4 - 5 293 130
BC068228.1 - 4 - 5 315 130

View Full Product Details