The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144541.1 | NP_653124.1 | 2 - 3 | 183 | 78 |
GenBank mRNA | AF440752.1 | - | 2 - 3 | 183 | 78 |
AK020047.1 | - | 2 - 3 | 269 | 78 | |
AK050695.1 | - | 2 - 3 | 228 | 78 | |
AK080991.1 | - | 2 - 3 | 219 | 78 | |
AK156929.1 | - | 2 - 3 | 275 | 78 | |
AK166884.1 | - | 2 - 3 | 219 | 78 | |
AK207416.1 | - | 2 - 3 | 182 | 78 | |
AK207670.1 | - | 2 - 3 | 182 | 78 | |
AK216664.1 | - | 1 - 2 | 189 | 78 | |
BC029277.1 | - | 2 - 3 | 534 | 78 | |
BC061000.1 | - | 2 - 3 | 259 | 78 |