Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Entpd5
Entrez Gene ID
12499
Gene Name
ectonucleoside triphosphate diphosphohydrolase 5
Gene Aliases
AI196558, AI987697, Cd39l4, ER-UDPase, NTPDase-5, NTPDase5, Pcph, mNTPase
Chromosome Location
Chr.12: 84373875 - 84409029 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026214.2 NP_001021385.1 7 - 8 789 80
NM_001286049.1 NP_001272978.1 6 - 7 764 80
NM_001286058.1 NP_001272987.1 5 - 6 691 80
NM_007647.3 NP_031673.2 6 - 7 716 80
GenBank mRNA AF006482.1 - 6 - 7 724 80
AF136571.1 - 6 - 7 750 80
AJ238636.1 - 6 - 7 752 80
AK002618.1 - 6 - 7 689 80
AK031581.1 - 7 - 8 769 80
AK036641.1 - 7 - 8 766 80
AK037736.1 - 6 - 7 685 80
AK045828.1 - 6 - 7 702 80
AK079267.1 - 5 - 6 613 80
AK080265.1 - 7 - 8 776 80
AK081435.1 - 7 - 8 754 80
AK088455.1 - 6 - 7 745 80
AK160950.1 - 4 - 5 610 80
AK163688.1 - 6 - 7 690 80
BC015247.1 - 5 - 6 674 80

View Full Product Details