Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd72
Entrez Gene ID
12517
Gene Name
CD72 antigen
Gene Aliases
CD72c, Ly-19, Ly-32, Ly-m19, Lyb-2
Chromosome Location
Chr.4: 43447724 - 43455468 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA J04170.1 - 7 - 8 946 87
S40777.1 - 7 - 8 963 87

View Full Product Details