The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301284.1 | NP_001288213.1 | 1 - 2 | 132 | 74 |
NM_007806.3 | NP_031832.2 | 1 - 2 | 132 | 74 | |
GenBank mRNA | AK018713.1 | - | 1 - 2 | 101 | 74 |
AK021200.1 | - | 1 - 2 | 120 | 74 | |
AK078854.1 | - | 1 - 2 | 103 | 74 | |
AK131580.1 | - | 1 - 2 | 101 | 74 | |
AK152414.1 | - | 1 - 2 | 123 | 74 | |
AK171748.1 | - | 1 - 2 | 101 | 74 | |
BC026791.1 | - | 1 - 2 | 83 | 74 | |
CF738398.1 | - | 1 - 2 | 115 | 74 | |
EU791539.1 | - | 1 - 2 | 62 | 74 | |
M31775.1 | - | 1 - 2 | 55 | 74 |