Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmo3
Entrez Gene ID
14262
Gene Name
flavin containing monooxygenase 3
Gene Aliases
AW111792
Chromosome Location
Chr.1: 162953800 - 162984491 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008030.1 NP_032056.1 8 - 9 1285 88
GenBank mRNA AK149440.1 - 8 - 9 1312 88
U87147.1 - 8 - 9 1285 88

View Full Product Details