The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009908.2 | NP_034038.2 | 4 - 5 | 772 | 124 |
GenBank mRNA | AJ006215.1 | - | 4 - 5 | 685 | 124 |
BC031500.1 | - | 4 - 5 | 701 | 124 | |
BC063776.1 | - | 4 - 5 | 701 | 124 |