The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271542.1 | NP_001258471.1 | 2 - 3 | 353 | 98 |
NM_001271543.1 | NP_001258472.1 | 2 - 3 | 424 | 98 | |
NM_011135.5 | NP_035265.1 | 2 - 3 | 424 | 98 | |
GenBank mRNA | AK007767.1 | - | 2 - 3 | 398 | 98 |
AK009561.1 | - | 2 - 3 | 412 | 98 | |
AK044535.1 | - | 1 - 2 | 1290 | 98 | |
AK132063.1 | - | 2 - 3 | 344 | 98 | |
AK137838.1 | - | 2 - 3 | 389 | 98 | |
AK147048.1 | - | 2 - 3 | 391 | 98 | |
AK166449.1 | - | 2 - 3 | 388 | 98 | |
BC006021.1 | - | 2 - 3 | 368 | 98 | |
CJ166755.1 | - | 2 - 3 | 424 | 98 |