The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013463.2 | NP_038491.2 | 1 - 2 | 269 | 64 |
GenBank mRNA | AK040081.1 | - | 1 - 2 | 201 | 64 |
AK054547.1 | - | 1 - 2 | 221 | 64 | |
AK145011.1 | - | 1 - 2 | 213 | 64 | |
AK164231.1 | - | 1 - 2 | 202 | 64 | |
AK166204.1 | - | 1 - 2 | 202 | 64 | |
AK166245.1 | - | 1 - 2 | 218 | 64 | |
BC009021.1 | - | 1 - 2 | 222 | 64 | |
U34071.1 | - | 1 - 2 | 219 | 64 | |
U50716.1 | - | 1 - 2 | 207 | 64 |