Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gtpbp1
Entrez Gene ID
14904
Gene Name
GTP binding protein 1
Gene Aliases
AL022987, AW045683, C85212, GP-1, Gtpbp
Chromosome Location
Chr.15: 79690896 - 79721479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013818.2 NP_038846.2 1 - 2 207 85
GenBank mRNA AK004612.1 - 1 - 2 208 85
AK147717.1 - 1 - 2 209 85
AK150068.1 - 1 - 2 214 85
AK150185.1 - 1 - 2 214 85
AK158342.1 - 1 - 2 213 85
AK170091.1 - 1 - 2 209 85
BC046228.1 - 1 - 2 207 85
U87965.1 - 1 - 2 94 85

View Full Product Details