Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gtpbp1
Entrez Gene ID
14904
Gene Name
GTP binding protein 1
Gene Aliases
AL022987, AW045683, C85212, GP-1, Gtpbp
Chromosome Location
Chr.15: 79690896 - 79721479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013818.2 NP_038846.2 9 - 10 1550 62
GenBank mRNA AK004612.1 - 9 - 10 1551 62
AK147717.1 - 9 - 10 1552 62
AK150068.1 - 9 - 10 1557 62
AK150185.1 - 9 - 10 1557 62
AK158342.1 - 9 - 10 1560 62
AK170091.1 - 9 - 10 1552 62
BC037129.1 - 7 - 8 1092 62
BC046228.1 - 9 - 10 1550 62
U87965.1 - 9 - 10 1437 62

View Full Product Details