Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Btg4
Entrez Gene ID
56057
Gene Name
B cell translocation gene 4
Gene Aliases
C86116, PC3B
Chromosome Location
Chr.9: 51102265 - 51119700 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019493.3 NP_062366.2 3 - 4 529 116
GenBank mRNA AB050983.2 - 3 - 4 529 116
AJ005120.1 - 3 - 4 402 116
AK136041.1 - 3 - 4 516 116
AK136124.1 - 3 - 4 517 116
AK136149.1 - 3 - 4 519 116
AK136185.1 - 3 - 4 521 116
AK161379.1 - 5 - 6 608 116
AK162122.1 - 3 - 4 516 116
AK163407.1 - 3 - 4 515 116
BC066811.1 - 3 - 4 503 116

View Full Product Details