Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clptm1
Entrez Gene ID
56457
Gene Name
cleft lip and palate associated transmembrane protein 1
Gene Aliases
HS9, N14
Chromosome Location
Chr.7: 19631580 - 19665030 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019649.2 NP_062623.2 3 - 4 337 63
GenBank mRNA AK152410.1 - 3 - 4 327 63
AK152809.1 - 3 - 4 327 63
AK165742.1 - 3 - 4 337 63
AK171901.1 - 3 - 4 328 63
BC022172.1 - 3 - 4 328 63
D67067.1 - 3 - 4 324 63

View Full Product Details