Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clptm1
Entrez Gene ID
56457
Gene Name
cleft lip and palate associated transmembrane protein 1
Gene Aliases
HS9, N14
Chromosome Location
Chr.7: 19631580 - 19665030 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019649.2 NP_062623.2 12 - 13 1582 67
GenBank mRNA AK148080.1 - 12 - 13 1564 67
AK165742.1 - 11 - 12 2276 67
BC018389.1 - 2 - 3 168 67
BC022172.1 - 12 - 13 1573 67
D67067.1 - 12 - 13 1568 67

View Full Product Details