Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ggcx
Entrez Gene ID
56316
Gene Name
gamma-glutamyl carboxylase
Gene Aliases
-
Chromosome Location
Chr.6: 72414296 - 72431106 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019802.5 NP_062776.1 1 - 2 142 77
GenBank mRNA AF087938.1 - 1 - 2 117 77
AK033496.1 - 1 - 2 69 77
AK036577.1 - 1 - 2 116 77
AK135425.1 - 1 - 2 129 77
AK165982.1 - 1 - 2 117 77
BC138763.1 - 1 - 2 67 77
BC138764.1 - 1 - 2 67 77
BY310874.1 - 1 - 2 142 77

View Full Product Details