The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021890.3 | NP_068690.3 | 7 - 8 | 1081 | 104 |
GenBank mRNA | AB041560.1 | - | 7 - 8 | 1023 | 104 |
AK029318.1 | - | 7 - 8 | 940 | 104 | |
AK080414.1 | - | 7 - 8 | 933 | 104 | |
AK090042.1 | - | 7 - 8 | 1064 | 104 | |
AK154534.1 | - | 7 - 8 | 958 | 104 | |
AK172509.1 | - | 7 - 8 | 1060 | 104 | |
BC055950.1 | - | 7 - 8 | 762 | 104 | |
BC071266.1 | - | 6 - 7 | 1069 | 104 |