The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163434.1 | NP_001156906.1 | 3 - 4 | 505 | 83 |
NM_022310.3 | NP_071705.3 | 2 - 3 | 587 | 83 | |
GenBank mRNA | AJ002387.1 | - | 2 - 3 | 366 | 83 |
AK004578.1 | - | 2 - 3 | 582 | 83 | |
AK076079.1 | - | 3 - 4 | 501 | 83 | |
AK077463.1 | - | 3 - 4 | 505 | 83 | |
AK144650.1 | - | 3 - 4 | 501 | 83 | |
AK144742.1 | - | 3 - 4 | 501 | 83 | |
AK146647.1 | - | 3 - 4 | 504 | 83 | |
AK148539.1 | - | 3 - 4 | 504 | 83 | |
AK149447.1 | - | 3 - 4 | 504 | 83 | |
AK151244.1 | - | 2 - 3 | 453 | 83 | |
AK151647.1 | - | 3 - 4 | 503 | 83 | |
AK151806.1 | - | 3 - 4 | 504 | 83 | |
AK152020.1 | - | 3 - 4 | 501 | 83 | |
AK152521.1 | - | 2 - 3 | 584 | 83 | |
AK152958.1 | - | 3 - 4 | 504 | 83 | |
AK159719.1 | - | 3 - 4 | 501 | 83 | |
AK160821.1 | - | - | 505 | 83 | |
AK166207.1 | - | 3 - 4 | 501 | 83 | |
AK166739.1 | - | 3 - 4 | 501 | 83 | |
AK167011.1 | - | 3 - 4 | 501 | 83 | |
AK168012.1 | - | 3 - 4 | 505 | 83 | |
AK168230.1 | - | 3 - 4 | 500 | 83 | |
AK169034.1 | - | 3 - 4 | 505 | 83 | |
AK192496.1 | - | 1 - 2 | 96 | 83 | |
AK197130.1 | - | 1 - 2 | 52 | 83 | |
AK198934.1 | - | 1 - 2 | 60 | 83 | |
BC005785.1 | - | 1 - 2 | 221 | 83 | |
BC050927.1 | - | 2 - 3 | 564 | 83 | |
D78645.1 | - | 2 - 3 | 447 | 83 |