The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026373.4 | NP_080649.1 | 2 - 3 | 254 | 63 |
GenBank mRNA | AK003686.2 | - | 2 - 3 | 220 | 63 |
AK012121.1 | - | 2 - 3 | 257 | 63 | |
AK018035.1 | - | 2 - 3 | 254 | 63 | |
BC025656.1 | - | 2 - 3 | 220 | 63 |