Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brms1l
Entrez Gene ID
52592
Gene Name
breast cancer metastasis-suppressor 1-like
Gene Aliases
0710008O11Rik, AI159718, BRMS1, D12Ertd407e
Chromosome Location
Chr.12: 55836243 - 55869736 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037756.2 NP_001032845.1 4 - 5 578 82
GenBank mRNA AK003055.1 - 4 - 5 327 82
AK154678.1 - 4 - 5 579 82
AK155742.1 - 4 - 5 579 82

View Full Product Details