The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029936.2 | NP_084212.2 | 15 - 16 | 2370 | 97 |
GenBank mRNA | AK019495.1 | - | 15 - 16 | 2350 | 97 |
AK166787.1 | - | 3 - 4 | 569 | 97 | |
BC049261.1 | - | 15 - 16 | 2311 | 97 | |
BC080729.1 | - | 15 - 16 | 2338 | 97 |