The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030260.3 | NP_084536.2 | 4 - 5 | 1434 | 64 |
NM_173002.3 | NP_766590.1 | 4 - 5 | 1434 | 64 | |
GenBank mRNA | AK030287.1 | - | 4 - 5 | 1350 | 64 |
AK042190.1 | - | 4 - 5 | 1361 | 64 | |
AK042392.1 | - | 4 - 5 | 1344 | 64 | |
AK044429.1 | - | 4 - 5 | 1327 | 64 | |
AK044529.1 | - | 4 - 5 | 1358 | 64 | |
AK044948.1 | - | 4 - 5 | 1328 | 64 | |
AK047079.1 | - | 4 - 5 | 1316 | 64 | |
AK047538.1 | - | 4 - 5 | 1350 | 64 | |
AK080307.1 | - | 4 - 5 | 1328 | 64 | |
AK081005.1 | - | 4 - 5 | 1436 | 64 | |
AK169517.1 | - | 4 - 5 | 64 | ||
AK205036.1 | - | 1 - 2 | 111 | 64 | |
BC003332.1 | - | 4 - 5 | 1418 | 64 |