The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053272.2 | NP_444502.2 | 1 - 2 | 362 | 51 |
GenBank mRNA | AK083737.1 | - | 1 - 2 | 317 | 51 |
AK129036.1 | - | 1 - 2 | 362 | 51 | |
AY039762.1 | - | 1 - 2 | 250 | 51 | |
BC019797.1 | - | 1 - 2 | 306 | 51 |