Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eps8l2
Entrez Gene ID
98845
Gene Name
EPS8-like 2
Gene Aliases
AI042819, AW545405, Eps8r2
Chromosome Location
Chr.7: 141338911 - 141363016 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133191.2 NP_573454.2 12 - 13 1157 80
GenBank mRNA BC005492.1 - 12 - 13 1157 80

View Full Product Details