The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133662.2 | NP_598423.1 | 1 - 2 | 217 | 73 |
GenBank mRNA | AK003466.1 | - | 1 - 2 | 243 | 72 |
AK051003.1 | - | 1 - 2 | 247 | 73 | |
AK170477.1 | - | 1 - 2 | 247 | 73 | |
BC006950.1 | - | 1 - 2 | 217 | 73 | |
CT010247.1 | - | 1 - 2 | 211 | 73 | |
CT010271.1 | - | 1 - 2 | 211 | 73 |