Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Osbpl9
Entrez Gene ID
100273
Gene Name
oxysterol binding protein-like 9
Gene Aliases
2600011I06Rik, AU015843, Orp-9
Chromosome Location
Chr.4: 109061145 - 109202272 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134791.2 NP_001128263.1 2 - 3 236 117
NM_133885.4 NP_598646.4 5 - 6 469 117
NM_173350.3 NP_775485.2 2 - 3 236 117
GenBank mRNA AK035658.1 - 2 - 3 185 117
AK131984.1 - 5 - 6 440 117
BC021507.1 - 1 - 2 97 117
BC023759.1 - 5 - 6 424 117
BC026927.1 - 2 - 3 119 117
BC089163.1 - 2 - 3 177 117
CJ088630.1 - 2 - 3 237 117

View Full Product Details