Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Osbpl9
Entrez Gene ID
100273
Gene Name
oxysterol binding protein-like 9
Gene Aliases
2600011I06Rik, AU015843, Orp-9
Chromosome Location
Chr.4: 109061145 - 109202272 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134791.2 NP_001128263.1 10 - 11 954 132
NM_133885.4 NP_598646.4 13 - 14 1187 132
NM_173350.3 NP_775485.2 11 - 12 993 132
GenBank mRNA AK035658.1 - 11 - 12 942 132
AK131984.1 - 13 - 14 1158 132
AK172010.1 - 11 - 12 980 132
BC021507.1 - 9 - 10 815 132
BC023759.1 - 13 - 14 1142 132
BC026927.1 - 11 - 12 876 132
BC089163.1 - 10 - 11 895 132

View Full Product Details