Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc22
Entrez Gene ID
54638
Gene Name
coiled-coil domain containing 22
Gene Aliases
AI481216, DXImx40e, Ppp1r3f, Sfc22
Chromosome Location
Chr.X: 7593809 - 7605420 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138603.3 NP_613069.3 7 - 8 1036 91
GenBank mRNA AF229637.1 - 7 - 8 977 91
AK010536.1 - 7 - 8 954 91
AK039573.1 - 7 - 8 1009 91
BC011195.1 - 7 - 8 941 91

View Full Product Details